siRNA Details
| Gene Symbol: TMEM33 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018126.2 | NP_060596.2 | 6 | 912 | ||
| XM_005248116.3 | XP_005248173.1 | 7 | 751 | |||
| XM_005248117.2 | XP_005248174.1 | 7 | 699 | |||
| GenBank mRNA | AK001387.1 | - | 6 | 880 | ||
| AK023062.1 | - | 6 | 892 | |||
| AK094190.1 | - | 7 | 823 | |||
| AK308501.1 | - | 5 | 692 | |||
| AY659966.1 | - | 6 | 880 | |||
| BC000948.2 | - | 6 | 642 | 3448888 | ||
| CR457260.1 | - | 6 | 547 | |||

TMEM33
Genomic Map