siRNA Details
| Gene Symbol: LRRC36 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018296.5 | NP_060766.5 | 3 | 378 | ||
| XM_005256025.2 | XP_005256082.1 | 3 | 378 | |||
| XM_005256026.2 | XP_005256083.1 | 3 | 378 | |||
| XM_005256027.2 | XP_005256084.1 | 3 | 378 | |||
| XM_005256028.1 | XP_005256085.1 | 2 | 250 | |||
| XM_011523200.1 | XP_011521502.1 | 3 | 378 | |||
| XM_011523203.1 | XP_011521505.1 | 3 | 378 | |||
| XM_011523205.1 | XP_011521507.1 | 3 | 357 | |||
| XM_011523206.2 | XP_011521508.1 | 3 | 378 | |||
| XM_017023400.1 | XP_016878889.1 | 3 | 378 | |||
| XM_017023401.1 | XP_016878890.1 | 3 | 378 | |||
| XM_017023402.1 | XP_016878891.1 | 3 | 378 | |||
| GenBank mRNA | AY963267.1 | - | 3 | 378 | ||
| BC026156.2 | - | 3 | 378 | 4828487 | ||

LRRC36
Genomic Map