siRNA Details
| Gene Symbol: CDCA7L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001127370.2 | NP_001120842.1 | 5 | 829 | ||
| NM_001127371.2 | NP_001120843.1 | 3 | 637 | |||
| NM_018719.4 | NP_061189.2 | 4 | 775 | |||
| GenBank mRNA | AK022955.1 | - | 4 | 750 | ||
| AK095965.1 | - | 5 | 783 | |||
| AK292490.1 | - | 4 | 750 | |||
| AK300781.1 | - | 3 | 597 | |||
| AY161168.1 | - | 4 | 652 | |||
| AY161169.1 | - | 4 | 655 | |||
| BC009352.2 | - | 4 | 667 | 4139111 | ||
| BC014630.2 | - | 3 | 619 | |||
| BC025242.1 | - | 4 | 729 | 4842169 | ||
| BC032576.1 | - | 4 | 740 | 5491780 | ||
| BC039823.1 | - | 4 | 739 | 6057601 | ||
| BX422040.2 | - | 3 | 585 | |||

CDCA7L
Genomic Map