siRNA Details
| Gene Symbol: TWSG1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020648.5 | NP_065699.1 | 5 | 730 | ||
| GenBank mRNA | AF196834.1 | - | 5 | 687 | ||
| AF294628.1 | - | 4 | 552 | |||
| AJ297391.1 | - | 5 | 681 | |||
| AK027629.1 | - | 7 | 888 | |||
| AK075550.1 | - | 5 | 730 | |||
| AK315751.1 | - | 5 | 615 | |||
| BC020490.1 | - | 5 | 644 | 3889103 | ||

TWSG1
Genomic Map