siRNA Details
| Gene Symbol: COG6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020751.2 | NP_065802.1 | 19 | 1973 | ||
| NR_026745.1 | 20 | 2038 | ||||
| XM_011535168.1 | XP_011533470.1 | 20 | 2058 | |||
| GenBank mRNA | AF116827.1 | - | 20 | 1921 | ||
| AK026638.1 | - | 18 | 1640 | |||
| AK294443.1 | - | 19 | 1973 | |||
| BC051723.1 | - | 19 | 1923 | 5263056 | ||
| CR627406.1 | - | 19 | 1916 | |||

COG6
Genomic Map