siRNA Details
| Gene Symbol: CCNB1IP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021178.4 | NP_067001.3 | 5 | 671 | ||
| NM_182849.2 | NP_878269.1 | 4 | 471 | |||
| NM_182852.3 | NP_878272.1 | 6 | 852 | |||
| GenBank mRNA | AF216381.1 | - | 1 | 182 | ||
| AK026233.1 | - | 5 | 638 | |||
| AK225253.1 | - | 5 | 643 | |||
| AK225451.1 | - | 5 | 638 | |||
| AK226037.1 | - | 5 | 643 | |||
| AK309183.1 | - | 5 | 640 | |||
| AL161994.1 | - | 5 | 651 | |||
| BC000369.2 | - | 4 | 613 | 2819346 | ||
| BC001218.1 | - | 4 | 616 | 3528496 | ||
| BC004435.1 | - | 4 | 613 | 2819346 | ||
| BI771433.1 | - | 5 | 654 | |||
| BI917081.1 | - | 5 | 662 | |||
| BM741413.1 | - | 4 | 438 | |||
| BX161423.1 | - | 5 | 831 | |||
| BX161424.1 | - | 5 | 799 | |||
| BX247978.1 | - | 5 | 779 | |||
| BX334446.2 | - | 5 | 794 | |||

CCNB1IP1
Genomic Map