siRNA Details
| Gene Symbol: HMHB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021182.1 | NP_067005.1 | 2 | 247 | ||
| GenBank mRNA | AF103884.1 | - | 2 | 247 | ||
| BC096711.1 | - | 2 | 181 | |||
| BC098244.1 | - | 2 | 181 | |||
| BC098251.2 | - | 2 | 181 | |||
| BC104412.1 | - | 2 | 181 | 40000018 | ||
| BC104413.1 | - | 2 | 181 | 40000019 | ||
| BC113894.1 | - | 2 | 181 | |||
| BC113971.1 | - | 2 | 181 | |||

HMHB1
Genomic Map