siRNA Details
| Gene Symbol: RPL38 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_000999.3 | NP_000990.1 | 4 | 260 | ||
| NM_001035258.1 | NP_001030335.1 | 4 | 225 | |||
| GenBank mRNA | AK312119.1 | - | 4 | 228 | ||
| BC000603.2 | - | 4 | 197 | 3346447 | ||
| BQ276548.1 | - | 4 | 256 | |||
| BU532782.1 | - | 4 | 228 | |||
| CB124619.1 | - | 4 | 250 | |||
| CB128529.1 | - | 4 | 203 | |||
| Z26876.1 | - | 4 | 242 | |||

RPL38
Genomic Map