siRNA Details
Gene Symbol: FAM229B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033564.2 | NP_001028736.1 | 4 | 1096 | ||
GenBank mRNA | BC044912.2 | - | 4 | 963 | 5286266 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: FAM229B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033564.2 | NP_001028736.1 | 4 | 1096 | ||
GenBank mRNA | BC044912.2 | - | 4 | 963 | 5286266 |