siRNA Details
| Gene Symbol: SDF2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_006923.3 | NP_008854.2 | 3 | 1190 | ||
| NR_045585.1 | 3 | 887 | ||||
| XM_011525106.2 | XP_011523408.1 | 5 | 1104 | |||
| GenBank mRNA | BC000500.2 | - | 3 | 905 | 2821329 | |
| BC001406.2 | - | 3 | 912 | 3140716 | ||

SDF2
Genomic Map