siRNA Details
| Gene Symbol: ECSCR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001077693.3 | NP_001071161.1 | 10 | 984 | ||
| NM_001293739.1 | NP_001280668.1 | 10 | 846 | |||
| NR_121659.1 | 11 | 955 | ||||
| GenBank mRNA | DQ462572.1 | - | 10 | 978 | ||

ECSCR
Genomic Map