siRNA Details
| Gene Symbol: NOC3L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022451.10 | NP_071896.8 | 14, 15 | 1791 | ||
| XM_017016544.1 | XP_016872033.1 | 12, 13 | 1844 | |||
| GenBank mRNA | AB077992.1 | - | 14, 15 | 1678 | ||
| AK022882.1 | - | 6, 7 | 702 | |||
| AK026953.1 | - | 4, 5 | 402 | |||
| AK091246.1 | - | 14, 15 | 1748 | |||
| AK292520.1 | - | 14, 15 | 1773 | |||
| AK302028.1 | - | 8, 9 | 876 | |||
| BC049850.1 | - | 14, 15 | 1741 | 5549517 | ||

NOC3L
Genomic Map