siRNA Details
| Gene Symbol: ERICH2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001290031.1 | NP_001276960.1 | 4 | 892 | ||
| NR_110644.1 | 5 | 1199 | ||||
| GenBank mRNA | BG215457.1 | - | 2 | 391 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ERICH2| Gene Symbol: ERICH2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001290031.1 | NP_001276960.1 | 4 | 892 | ||
| NR_110644.1 | 5 | 1199 | ||||
| GenBank mRNA | BG215457.1 | - | 2 | 391 | ||