siRNA Details
| Gene Symbol: LYPLAL1-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038845.1 | 2, 3 | 410 | |||
| GenBank mRNA | BC086863.1 | - | 2, 3 | 373 | ||
| DA132981.1 | - | 2, 3 | 410 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LYPLAL1-AS1| Gene Symbol: LYPLAL1-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038845.1 | 2, 3 | 410 | |||
| GenBank mRNA | BC086863.1 | - | 2, 3 | 373 | ||
| DA132981.1 | - | 2, 3 | 410 | |||