siRNA Details
| Gene Symbol: PCNX4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022495.5 | NP_071940.4 | 3 | 1001 | ||
| XM_005268009.2 | XP_005268066.1 | 4 | 2378 | |||
| GenBank mRNA | AK095489.1 | - | 3 | 985 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

PCNX4| Gene Symbol: PCNX4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022495.5 | NP_071940.4 | 3 | 1001 | ||
| XM_005268009.2 | XP_005268066.1 | 4 | 2378 | |||
| GenBank mRNA | AK095489.1 | - | 3 | 985 | ||