siRNA Details
| Gene Symbol: LOC101927139 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_123718.1 | 3 | 353 | |||
| GenBank mRNA | BX109047.1 | - | 3 | 353 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101927139| Gene Symbol: LOC101927139 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_123718.1 | 3 | 353 | |||
| GenBank mRNA | BX109047.1 | - | 3 | 353 | ||