siRNA Details
| Gene Symbol: MROH3P | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | DQ983818.1 | - | 16 | 2056 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

MROH3P| Gene Symbol: MROH3P | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | DQ983818.1 | - | 16 | 2056 | ||