siRNA Details
| Gene Symbol: LINC00273 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038368.1 | 1 | 1272 | |||
| GenBank mRNA | AY587847.1 | - | 1 | 1272 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC00273| Gene Symbol: LINC00273 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038368.1 | 1 | 1272 | |||
| GenBank mRNA | AY587847.1 | - | 1 | 1272 | ||