siRNA Details
| Gene Symbol: WNT7B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_058238.2 | NP_478679.1 | 4 | 1381 | ||
| XM_011530366.1 | XP_011528668.1 | 4 | 1146 | |||
| GenBank mRNA | AB062766.1 | - | 4 | 1012 | ||
| AF416743.1 | - | 4 | 1381 | |||
| AK123019.1 | - | 4 | 976 | |||
| AK289526.1 | - | 4 | 1146 | |||
| BC034923.1 | - | 4 | 1104 | 5457045 | ||

WNT7B
Genomic Map