siRNA Details
| Gene Symbol: MRPS24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032014.2 | NP_114403.1 | 4 | 663 | ||
| GenBank mRNA | AF161453.1 | - | 4 | 590 | ||
| BC012167.1 | - | 4 | 615 | 4637888 | ||
| BC054865.1 | - | 4 | 630 | 6456699 | ||
| Gene Symbol: URGCP-MRPS24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001204871.1 | NP_001191800.1 | 7 | 805 | ||

MRPS24
Genomic Map