siRNA Details
| Gene Symbol: LOC100996432 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_136571.1 | 2 | 131 | |||
| GenBank mRNA | DB073469.1 | - | 2 | 108 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100996432| Gene Symbol: LOC100996432 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_136571.1 | 2 | 131 | |||
| GenBank mRNA | DB073469.1 | - | 2 | 108 | ||