siRNA Details
| Gene Symbol: RNF208 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031297.4 | NP_112587.2 | 1 | 889 | ||
| GenBank mRNA | AF416715.1 | - | 1 | 570 | ||
| AL136540.1 | - | 1 | 572 | |||
| BC016958.1 | - | 1 | 578 | 4151712 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

RNF208| Gene Symbol: RNF208 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031297.4 | NP_112587.2 | 1 | 889 | ||
| GenBank mRNA | AF416715.1 | - | 1 | 570 | ||
| AL136540.1 | - | 1 | 572 | |||
| BC016958.1 | - | 1 | 578 | 4151712 | ||