siRNA Details
| Gene Symbol: PAXIP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007349.3 | NP_031375.3 | 8 | 1905 | ||
| XM_005249539.1 | XP_005249596.1 | 9 | 2044 | |||
| XM_011515982.2 | XP_011514284.1 | 8 | 2019 | |||
| XM_011515983.1 | XP_011514285.1 | 8 | 1905 | |||
| XM_017011881.1 | XP_016867370.1 | 9 | 3217 | |||
| GenBank mRNA | AK093584.1 | - | 2 | 599 | ||
| AK123044.1 | - | 4 | 1950 | |||
| AK123600.1 | - | 9 | 2044 | |||
| AK293744.1 | - | 6 | 1737 | |||
| AK307417.1 | - | 8 | 1905 | |||
| BC033781.1 | - | 3 | 1146 | 4509264 | ||
| BX538201.1 | - | 2 | 681 | |||
| BX640616.1 | - | 8 | 3803 | |||
| U80735.1 | - | 4 | 888 | |||

PAXIP1
Genomic Map