siRNA Details
| Gene Symbol: UBAC2-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_036531.1 | 2 | 576 | |||
| NR_036532.1 | 2 | 302 | ||||
| GenBank mRNA | AK097488.1 | - | 2 | 581 | ||
| BC043515.1 | - | 2 | 257 | |||
| DB446060.1 | - | 2 | 303 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

UBAC2-AS1| Gene Symbol: UBAC2-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_036531.1 | 2 | 576 | |||
| NR_036532.1 | 2 | 302 | ||||
| GenBank mRNA | AK097488.1 | - | 2 | 581 | ||
| BC043515.1 | - | 2 | 257 | |||
| DB446060.1 | - | 2 | 303 | |||