siRNA Details
| Gene Symbol: KLHL33 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109997.2 | NP_001103467.2 | 2 | 260 | ||
| XM_005267347.4 | XP_005267404.1 | 3 | 1259 | |||
| XM_011536450.2 | XP_011534752.1 | 3 | 975 | |||
| GenBank mRNA | BY797694.2 | - | 2 | 260 | ||

KLHL33
Genomic Map