siRNA Details
| Gene Symbol: TMEM116 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK021528.1 | - | 1 | 1126 | ||
| AK092832.1 | - | 1 | 2410 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

TMEM116| Gene Symbol: TMEM116 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK021528.1 | - | 1 | 1126 | ||
| AK092832.1 | - | 1 | 2410 | |||