siRNA Details
| Gene Symbol: OR7E154P | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK122996.1 | - | 1 | 4159 | ||
| Gene Symbol: OR7E125P | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
| GenBank mRNA | AF359416.1 | - | LIT2085 PJCG6 pseudogene |
1 | 236 | |

OR7E154P
Genomic Map