siRNA Details
| Gene Symbol: CROCC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_017005450.1 | XP_016860939.1 | 17, 18 | 2855 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

CROCC2| Gene Symbol: CROCC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_017005450.1 | XP_016860939.1 | 17, 18 | 2855 | ||