siRNA Details
| Gene Symbol: ACIN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001164814.1 | NP_001158286.1 | 6 | 1583 | ||
| NM_001164815.1 | NP_001158287.1 | 5 | 1463 | |||
| NM_014977.3 | NP_055792.1 | 6 | 1583 | |||
| XM_005267415.3 | XP_005267472.1 | 6 | 1387 | |||
| XM_005267416.3 | XP_005267473.1 | 6 | 1387 | |||
| XM_006720081.3 | XP_006720144.1 | 6 | 1387 | |||
| XM_011536570.2 | XP_011534872.1 | 6 | 1387 | |||
| GenBank mRNA | AB014570.2 | - | 6 | 1092 | ||
| AF124726.1 | - | 6 | 1582 | |||
| AK299034.1 | - | 5 | 968 | |||
| BC140805.1 | - | 6 | 1352 | 9021673 | ||

ACIN1
Genomic Map