siRNA Details
| Gene Symbol: LOC105379194 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_134244.1 | 1 | 121 | |||
| GenBank mRNA | HY038745.1 | - | 1 | 122 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC105379194| Gene Symbol: LOC105379194 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_134244.1 | 1 | 121 | |||
| GenBank mRNA | HY038745.1 | - | 1 | 122 | ||