siRNA Details
Gene Symbol: LOC101929243 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_134625.1 | 2 | 809 | |||
NR_134626.1 | 2 | 293 | ||||
GenBank mRNA | CB045917.1 | - | 2 | 302 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC101929243 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_134625.1 | 2 | 809 | |||
NR_134626.1 | 2 | 293 | ||||
GenBank mRNA | CB045917.1 | - | 2 | 302 |