siRNA Details
| Gene Symbol: PCNX1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_014982.2 | NP_055797.2 | 11 | 3247 | ||
| XM_005267420.4 | XP_005267477.1 | 11 | 3209 | |||
| XM_005267421.4 | XP_005267478.1 | 11 | 3206 | |||
| XM_005267424.3 | XP_005267481.2 | 11 | 2913 | |||
| XM_005267425.4 | XP_005267482.1 | 9 | 2891 | |||
| XM_005267427.4 | XP_005267484.1 | 11 | 3209 | |||
| XM_005267428.3 | XP_005267485.1 | 4 | 197 | |||
| XM_017021096.1 | XP_016876585.1 | 9 | 2891 | |||
| GenBank mRNA | AF233450.1 | - | 11 | 3247 | ||

PCNX1
Genomic Map