siRNA Details
| Gene Symbol: C3orf58 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173552.3 | NP_775823.1 | 1 | 246 | ||
| XM_011512543.2 | XP_011510845.1 | 1 | 135 | |||
| GenBank mRNA | AK315082.1 | - | 1 | 220 | ||
| DA760601.1 | - | 1 | 245 | |||
| DB067725.1 | - | 1 | 247 | |||

C3orf58
Genomic Map