siRNA Details
| Gene Symbol: LOC101928881 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_136325.1 | 3 | 576 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101928881| Gene Symbol: LOC101928881 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_136325.1 | 3 | 576 | |||