siRNA Details
| Gene Symbol: SHISA9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001145204.2 | NP_001138676.2 | 5 | 1355 | ||
| XM_005255539.3 | XP_005255596.2 | 4 | 1308 | |||
| GenBank mRNA | AK299610.1 | - | 6 | 785 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

SHISA9| Gene Symbol: SHISA9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001145204.2 | NP_001138676.2 | 5 | 1355 | ||
| XM_005255539.3 | XP_005255596.2 | 4 | 1308 | |||
| GenBank mRNA | AK299610.1 | - | 6 | 785 | ||