siRNA Details
| Gene Symbol: CEP152 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001194998.1 | NP_001181927.1 | 27 | 4761 | ||
| NM_014985.3 | NP_055800.2 | 26 | 4593 | |||
| XM_006720437.3 | XP_006720500.1 | 27 | 4946 | |||
| XM_011521373.2 | XP_011519675.1 | 27 | 5484 | |||
| XM_011521379.2 | XP_011519681.1 | 26 | 5408 | |||
| XM_011521381.2 | XP_011519683.1 | 15 | 3674 | |||
| XM_017022014.1 | XP_016877503.1 | 20 | 3707 | |||
| XM_017022015.1 | XP_016877504.1 | 15 | 2863 | |||
| GenBank mRNA | BC117182.1 | - | 26 | 4434 | 40125733 | |
| BX648822.1 | - | 27 | 4688 | |||

CEP152
Genomic Map