siRNA Details
Gene Symbol: DIO3OS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF469199.1 | - | 2 | 173 | ||
AF469200.1 | - | 2 | 173 | |||
AF469204.1 | - | 1 | 21 | |||
BC065701.1 | - | 3 | 375 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: DIO3OS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF469199.1 | - | 2 | 173 | ||
AF469200.1 | - | 2 | 173 | |||
AF469204.1 | - | 1 | 21 | |||
BC065701.1 | - | 3 | 375 |