siRNA Details
| Gene Symbol: CCDC28B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001301011.1 | NP_001287940.1 | 5 | 1567 | ||
| NM_024296.4 | NP_077272.2 | 6 | 994 | |||
| XM_011542113.2 | XP_011540415.1 | 5 | 2326 | |||
| XM_017002307.1 | XP_016857796.1 | 6 | 1748 | |||
| XM_017002308.1 | XP_016857797.1 | 5 | 800 | |||
| XM_017002309.1 | XP_016857798.1 | 5 | 1557 | |||
| GenBank mRNA | AK225113.1 | - | 6 | 764 | ||
| AK291904.1 | - | 6 | 1004 | |||
| BC022848.2 | - | 6 | 728 | 3946778 | ||

CCDC28B
Genomic Map