siRNA Details
| Gene Symbol: NSUN7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024677.4 | NP_078953.3 | 10 | 1882 | ||
| XM_017008611.1 | XP_016864100.1 | 9 | 1761 | |||
| XM_017008612.1 | XP_016864101.1 | 8 | 1659 | |||
| XM_017008613.1 | XP_016864102.1 | 10 | 1905 | |||
| XM_017008614.1 | XP_016864103.1 | 10 | 1905 | |||
| GenBank mRNA | AK094298.1 | - | 2 | 122 | ||
| AK304048.1 | - | 10 | 1606 | |||
| BC036568.2 | - | 10 | 1854 | 5272990 | ||
| BC042401.1 | - | 10 | 1882 | |||

NSUN7
Genomic Map