siRNA Details
| Gene Symbol: ASB13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024701.3 | NP_078977.2 | 5 | 638 | ||
| NR_024581.1 | 4 | 487 | ||||
| NR_037164.1 | 6 | 719 | ||||
| GenBank mRNA | AF403031.1 | - | 5 | 620 | ||
| AK023196.1 | - | 5 | 621 | |||
| AK091935.1 | - | 4 | 487 | |||
| AK292071.1 | - | 5 | 624 | |||
| BC012056.1 | - | 6 | 696 | 4552643 | ||
| CR457302.1 | - | 5 | 594 | |||

ASB13
Genomic Map