siRNA Details
| Gene Symbol: C10orf95 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024886.2 | NP_079162.1 | 1, 2 | 156 | ||
| GenBank mRNA | AK024342.1 | - | 1, 2 | 156 | ||
| BC126459.1 | - | 1, 2 | 101 | 8992175 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

C10orf95| Gene Symbol: C10orf95 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024886.2 | NP_079162.1 | 1, 2 | 156 | ||
| GenBank mRNA | AK024342.1 | - | 1, 2 | 156 | ||
| BC126459.1 | - | 1, 2 | 101 | 8992175 | ||