siRNA Details
| Gene Symbol: ESRP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024939.2 | NP_079215.2 | 13 | 1962 | ||
| XM_005256153.4 | XP_005256210.1 | 13 | 1901 | |||
| XM_006721273.3 | XP_006721336.1 | 13 | 1970 | |||
| XM_011523338.2 | XP_011521640.1 | 12 | 1862 | |||
| GenBank mRNA | AF289577.1 | - | 7 | 1064 | ||
| AK025571.1 | - | 13 | 1891 | |||
| AK025901.1 | - | 2 | 227 | |||
| BC030146.2 | - | 13 | 1784 | 4810948 | ||
| BC069241.1 | - | 13 | 2313 | |||
| LN680438.1 | - | 13 | 1774 | |||
| LN680439.1 | - | 13 | 1774 | |||

ESRP2
Genomic Map