siRNA Details
| Gene Symbol: CCDC92 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001304957.1 | NP_001291886.1 | 5 | 800 | ||
| NM_001304958.1 | NP_001291887.1 | 6 | 907 | |||
| NM_001304959.1 | NP_001291888.1 | 5 | 549 | |||
| NM_001304960.1 | NP_001291889.1 | 7 | 718 | |||
| NM_001304961.1 | NP_001291890.1 | 4 | 717 | |||
| NM_025140.2 | NP_079416.1 | 5 | 810 | |||
| XM_005253624.2 | XP_005253681.1 | 5 | 554 | |||
| XM_017019984.1 | XP_016875473.1 | 5 | 1368 | |||
| GenBank mRNA | AB015292.1 | - | 4 | 330 | ||
| AK026124.1 | - | 5 | 441 | |||
| AK054680.1 | - | 3 | 2077 | |||
| AK124358.1 | - | 1 | 1899 | |||
| AK125866.1 | - | 3 | 2328 | |||
| AK222661.1 | - | 5 | 398 | |||
| AW953473.1 | - | 8 | 689 | |||
| BC017186.2 | - | 5 | 433 | 3051346 | ||
| BX394686.2 | - | 4 | 466 | |||
| DA370549.1 | - | 6 | 505 | |||

CCDC92
Genomic Map