siRNA Details
| Gene Symbol: LOC81787 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AL512723.1 | - | 1 | 966 | ||
| Gene Symbol: LOC107987377 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_017030113.1 | XP_016885602.1 | 4 | 1570 | ||

LOC81787
Genomic Map