siRNA Details
| Gene Symbol: SPEN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015001.2 | NP_055816.2 | 11 | 3707 | ||
| GenBank mRNA | AF356524.1 | - | 11 | 3707 | ||
| AK000882.1 | - | 1 | 520 | |||
| AK022949.1 | - | 1 | 1128 | |||
| AK092332.1 | - | 7 | 2280 | |||
| AL096858.1 | - | 8 | 2625 | |||

SPEN
Genomic Map