siRNA Details
| Gene Symbol: TMUB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001136044.1 | NP_001129516.1 | 2 | 447 | ||
| NM_031434.3 | NP_113622.1 | 2 | 910 | |||
| GenBank mRNA | AK314035.1 | - | 2 | 675 | ||
| AY037155.1 | - | 2 | 428 | |||
| AY358481.1 | - | 2 | 406 | |||
| AY603380.1 | - | 1 | 432 | |||
| BC000936.1 | - | 2 | 447 | 3449979 | ||
| BC033182.1 | - | 2 | 585 | 4586932 | ||

TMUB1
Genomic Map