siRNA Details
| Gene Symbol: C11orf68 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001135635.1 | NP_001129107.1 | 2 | 749 | ||
| NM_031450.3 | NP_113638.2 | 2 | 746 | |||
| GenBank mRNA | AF073483.1 | - | 2 | 655 | ||
| BC004414.1 | - | 1 | 573 | |||
| BC010512.1 | - | 1 | 740 | 4150027 | ||
| BC013185.1 | - | 2 | 746 | 4346483 | ||

C11orf68
Genomic Map