siRNA Details
| Gene Symbol: HIST1H4B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_003544.2 | NP_003535.1 | 1 | 313 | ||
| GenBank mRNA | BC069467.1 | - | 1 | 313 | 7262716 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

HIST1H4B| Gene Symbol: HIST1H4B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_003544.2 | NP_003535.1 | 1 | 313 | ||
| GenBank mRNA | BC069467.1 | - | 1 | 313 | 7262716 | |