siRNA Details
| Gene Symbol: KRTAP17-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031964.1 | NP_114170.1 | 1 | 644 | ||
| GenBank mRNA | AJ406952.1 | - | 1 | 644 | ||
| BC069078.1 | - | 1 | 642 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

KRTAP17-1| Gene Symbol: KRTAP17-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031964.1 | NP_114170.1 | 1 | 644 | ||
| GenBank mRNA | AJ406952.1 | - | 1 | 644 | ||
| BC069078.1 | - | 1 | 642 | |||