siRNA Details
| Gene Symbol: TM2D1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032027.2 | NP_114416.1 | 3 | 569 | ||
| NR_135160.1 | 3 | 569 | ||||
| NR_135161.1 | 3 | 569 | ||||
| NR_135162.1 | 3 | 592 | ||||
| GenBank mRNA | AF353990.1 | - | 3 | 569 | ||
| AI168073.1 | - | 3 | 263 | |||
| AK311444.1 | - | 3 | 215 | |||
| AK311705.1 | - | 3 | 216 | |||
| BC029486.1 | - | 3 | 287 | 5271098 | ||
| BG700625.1 | - | 3 | 284 | |||
| BU664262.1 | - | 3 | 236 | |||

TM2D1
Genomic Map